Learn More
Cayman Chemical 8Isoprostn AffInIty Column 1ea
SDP
Catalog No. 502942337
Encompass_Preferred
Cayman’s 8-Isoprostane Affinity Column provides a specific and convenient means for the purification of 8-isoprostane (8-iso PGF2α) from urine and plasma samples. EAch column contains 0.5 ml of affinity sorbent (mouse anti-8-isoprostane monoclonal antibody covalently bound to Sepharose 4).
Catalog No. 50-294-2337 Supplier Cayman Chemical Supplier No. 4011111 EA
Cayman’s 8-Isoprostane Affinity Column provides a specific and convenient means for the purification of 8-isoprostane (8-iso PGF2α) from urine and plasma samples. EAch column contains 0.5 ml of affinity sorbent (mouse anti-8-isoprostane monoclonal antibody covalently bound to Sepharose 4).