Learn More
Cayman Chemical 8Isoprostn AffInIty Sorben 1mL
SDP
Catalog No. 502942495
Encompass_Preferred
Cayman’s 8-Isoprostane Affinity Sorbent provides a specific and convenient means for the purification of 8-isoprostane (8-iso PGF2α) from urine and plasma samples. EAch vial contains 1 ml of affinity sorbent (mouse anti-8-isoprostane monoclonal antibody covalently bound to Sepharose 4).
Catalog No. 50-294-2495 Supplier Cayman Chemical Supplier No. 4011131 ML
Cayman’s 8-Isoprostane Affinity Sorbent provides a specific and convenient means for the purification of 8-isoprostane (8-iso PGF2α) from urine and plasma samples. EAch vial contains 1 ml of affinity sorbent (mouse anti-8-isoprostane monoclonal antibody covalently bound to Sepharose 4).