Learn More
Cayman Chemical 8IsoTetrahydrocnabIvarIn 100ug
SDP
Catalog No. 502942931
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in hashish oil; intended for research and forensic applications
Catalog No. 50-294-2931 Supplier Cayman Chemical Supplier No. 40474100 UG
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in hashish oil; intended for research and forensic applications