Learn More
Cole-Parmer Techne FBATH08 Unheated Water Bath, Stainless Steel, 8L
Supplier:  Cole-Parmer 1610453
Encompass
Designed to be used with a clip-on Tempette or Tempunit thermoregulator (sold separately), these water baths incorporate carrying handles for added safety. All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.
Catalog No. 50-196-0475