Learn More
Teknova 8M Guanidine HCl, 50mM Tris HCl. 1000mL, Non-Sterile.
SDP
Catalog No. NC1028875
Encompass
8.0M Guanidine-HCl, 50mM Tris-HCl,0.062% Sodium Hydroxide
Catalog No. NC1028875 Supplier Teknova Supplier No. G0325
8.0M Guanidine-HCl, 50mM Tris-HCl,0.062% Sodium Hydroxide