Learn More
Cayman Chemical 8-Methoxymethyl-3-isobutyl-1-methylxanthine, Purity: ≥98%, CAS: 78033-08-6, 5 mg
SDP
Catalog No. 502918346
Encompass_Preferred
A PDE1 inhibitor (IC50 = 5 µM); selective for PDE1 over PDE2, PDE3, and PDE4 but also inhibits PDE5 (IC50s = 37, >400, >400, and 2 µM, respectively); enhances forskolin-induced increases in intracellular calcium levels in isolated rat olfactory receptor neurons at 30 µM; induces relaxation of isolated human saphenous vein segments in the absence of endothelium (EC50 = 31 µM); enhances iloprost-induced decreases in pulmonary artery pressure and vascular resistance in a rabbit model of U-46619-induced pulmonary hypertension at 200 µg/kg
Catalog No. 50-291-8346 Supplier Cayman Chemical Supplier No. 391515 MG
A PDE1 inhibitor (IC50 = 5 µM); selective for PDE1 over PDE2, PDE3, and PDE4 but also inhibits PDE5 (IC50s = 37, >400, >400, and 2 µM, respectively); enhances forskolin-induced increases in intracellular calcium levels in isolated rat olfactory receptor neurons at 30 µM; induces relaxation of isolated human saphenous vein segments in the absence of endothelium (EC50 = 31 µM); enhances iloprost-induced decreases in pulmonary artery pressure and vascular resistance in a rabbit model of U-46619-induced pulmonary hypertension at 200 µg/kg