Learn More
S2 Media 8mL Brain Heart Infusion (BHI) Agar with 7% Sheep Blood Slants, 20x125mm PC Tubes, 50/box
Catalog No. 502995888
Encompass_Preferred
Brain Heart Infusion (BHI) Agar with 7% Sheep Blood is an enriched, non-selective, and differential medium is used for the isolation and cultivation of a wide variety of fungal species, including systemic fungi. This medium was first used for the isolation of dental pathogens. Phosphate buffer acts to neutralize the acids produced from the utilization of glucose and thus maintain viability. • Product meets AOAC, BAM, CLSI, EPA, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• BHI with 7% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5888 Supplier S2 Media Supplier No. 5509
Brain Heart Infusion (BHI) Agar with 7% Sheep Blood is an enriched, non-selective, and differential medium is used for the isolation and cultivation of a wide variety of fungal species, including systemic fungi. This medium was first used for the isolation of dental pathogens. Phosphate buffer acts to neutralize the acids produced from the utilization of glucose and thus maintain viability. • Product meets AOAC, BAM, CLSI, EPA, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• BHI with 7% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.