Learn More
Foxx Life Sciences 8mL Narrow Mouth HDPE Amber Reagent Bottles, 72CS
GREENER_CHOICE
Catalog No. 5000846712
Encompass_Preferred
EZLabpure™  8mL HDPE Amber Reagent Bottles provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth HDPE Reagent Bottle design ensures precision dispensing, controlled pouring, and minimal spillage. Amber HDPE chemical resistant reagent bottles block UV radiation to protect light-sensitive chemicals, ensuring safe, long-term storage in pharmaceutical labs.
Features
HDPE Amber Bottles with UV-blocking opaque color for light-sensitive samples.
Leak-proof Linerless screw caps for secure sealing.
Lightweight and durable, HDPE storage reagent bottles are designed for long-term storage.
HDPE Amber Bottles with Low Liquid Retention
Catalog No. 50-008-46712 Supplier Foxx Life Sciences Supplier No. 76K1311FLS
EZLabpure™  8mL HDPE Amber Reagent Bottles provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth HDPE Reagent Bottle design ensures precision dispensing, controlled pouring, and minimal spillage. Amber HDPE chemical resistant reagent bottles block UV radiation to protect light-sensitive chemicals, ensuring safe, long-term storage in pharmaceutical labs.
Features
HDPE Amber Bottles with UV-blocking opaque color for light-sensitive samples.
Leak-proof Linerless screw caps for secure sealing.
Lightweight and durable, HDPE storage reagent bottles are designed for long-term storage.
HDPE Amber Bottles with Low Liquid Retention