Learn More
Analytical Sales & Services 8mm, 100µL, 12 x 32mm Clear Glass Screw Top MicroVial
Catalog No. 5000804487
Encompass
Advantage™ 8mm Clear Glass 100µL Screw Top MicroVial
Catalog No. 50-008-04487 Supplier Analytical Sales & Services Supplier No. 21203
Advantage™ 8mm Clear Glass 100µL Screw Top MicroVial