Learn More
Analytical Sales & Services 8mm, 2mL, 12 x 32 Amber Glass Screw Vial - Silanized
Catalog No. 5000804223
Encompass
Advantage™ 8mm, 2mL Volume, 12 x 32 Standard Amber Glass Screw Vial - Silanized
Catalog No. 50-008-04223 Supplier Analytical Sales & Services Supplier No. 21232SCASE
Advantage™ 8mm, 2mL Volume, 12 x 32 Standard Amber Glass Screw Vial - Silanized