Learn More
Chemglass Life Sciences Glassblowers Condenser Coil, 8mm OD, Coil Dimension 28mm OD X 280mm Length
SDP
Catalog No. CG62702
Encompass_Preferred
Glassblowers Condenser Coil, 8mm OD, Coil Dimension 28mm OD X 280mm Length. For use in construction of spiral or Graham style condensers. Coils can be sealed together to form longer lengths.
Catalog No. CG62702 Supplier Chemglass Life Sciences Supplier No. CG62702
Glassblowers Condenser Coil, 8mm OD, Coil Dimension 28mm OD X 280mm Length. For use in construction of spiral or Graham style condensers. Coils can be sealed together to form longer lengths.