Learn More
Across International 8mm Diameter ID Hardened Steel Dry Pressing Die Set
SDP
Catalog No. 502251344
Encompass_Preferred
A set of 8mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 8mm diameter cylinder-shape pellets.
Catalog No. 50-225-1344 Supplier Across International Supplier No. SDS8
A set of 8mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 8mm diameter cylinder-shape pellets.