Learn More
TA Instruments 8mm Focusing Lens, Designed for LED Heads, Precision Optical Accessory, Concentrates Light Output, Enhances Spot Size Control and Application Efficiency
Catalog No. 5000580869 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments 8mm Focusing Lens is designed for use with OmniCure LED Heads to optimize light delivery. This precision optical accessory focuses and concentrates the LED output to a smaller spot size, improving curing efficiency and consistency. By narrowing and controlling the beam, the 8mm lens enables enhanced accuracy in applications requiring high precision. It is an essential accessory for users looking to maximize light intensity while maintaining controlled curing areas, particularly in specialized industrial and research environments.
Catalog No. 50-005-80869 Supplier TA Instruments Supplier No. 206440.001
Waters TA Instruments 8mm Focusing Lens is designed for use with OmniCure LED Heads to optimize light delivery. This precision optical accessory focuses and concentrates the LED output to a smaller spot size, improving curing efficiency and consistency. By narrowing and controlling the beam, the 8mm lens enables enhanced accuracy in applications requiring high precision. It is an essential accessory for users looking to maximize light intensity while maintaining controlled curing areas, particularly in specialized industrial and research environments.