Learn More
Ace Glass, Inc. 8mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass
SDP
Supplier:  Ace Glass, Inc. 845512
Encompass
8mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-593