Learn More
Chemglass Life Sciences Tube, Connecting, "Y" Shaped, CG-300-01 (9mm Small Serration, 10mm Large Serration) Hose Connection
SDP
Catalog No. CG402301
Encompass_Preferred
Tube, Connecting, "Y" Shaped, CG-300-01 (9mm Small Serration, 10mm Large Serration) Hose Connection. Y shaped tube having serrated ends for connection of rubber, latex or Tygon tubing.
Catalog No. CG402301 Supplier Chemglass Life Sciences Supplier No. CG402301
Tube, Connecting, "Y" Shaped, CG-300-01 (9mm Small Serration, 10mm Large Serration) Hose Connection. Y shaped tube having serrated ends for connection of rubber, latex or Tygon tubing.