Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Plug, 8mm Bore, 24/40 Plug Size, 16mm OD Sidearms
SDP
Catalog No. CG42408
Encompass_Preferred
Stopcock, Straight Bore, PTFE Plug, 8mm Bore, 24/40 Plug Size, 16mm OD Sidearms.Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. CG42408 Supplier Chemglass Life Sciences Supplier No. CG42408
Stopcock, Straight Bore, PTFE Plug, 8mm Bore, 24/40 Plug Size, 16mm OD Sidearms.Straight bore stopcock having a 1:5 taper solid PTFE plug.