Learn More
Chemglass Life Sciences 8mm Shaft Dia. Applikon Agitator to IKA HA.mt.s.10 Big Motor Adapter Kit
SDP
Catalog No. 5000980643
Encompass
Agitator Adapter to adapt IKA HABITAT motors to Applikon style agitator assemblies. Small motor adapters are for use with 1L to 5L (8mm Shaft) bioreactors and the big motor adapters are for use with 7L (8mm shaft) and 15L (8mm shaft or 15mm) agitator assemblies.Note: This is the adapter kit only and does not include the agitator assembly. For the 8mm agitator assemblies’ removal of the (4) rods and upper motor support portion is required. The adapter screws into the mounting holes used by the rods. For the 15mm agitator assembly (4) existing screws on the top plate will need to be removed and the adapter will screw in using these holes.
Catalog No. 50-009-80643 Supplier Chemglass Life Sciences Supplier No. CLS1380IKA10
Agitator Adapter to adapt IKA HABITAT motors to Applikon style agitator assemblies. Small motor adapters are for use with 1L to 5L (8mm Shaft) bioreactors and the big motor adapters are for use with 7L (8mm shaft) and 15L (8mm shaft or 15mm) agitator assemblies.Note: This is the adapter kit only and does not include the agitator assembly. For the 8mm agitator assemblies’ removal of the (4) rods and upper motor support portion is required. The adapter screws into the mounting holes used by the rods. For the 15mm agitator assembly (4) existing screws on the top plate will need to be removed and the adapter will screw in using these holes.