Learn More
Beaver Visitec Intl Ltd TREPHINE BLADE, 8.0MM, STERILE, 1/EA
Catalog No. NC1312079
Encompass_Preferred
Beaver-Visitec o ers a comprehensive selection of corneal trephine blades, handles and tissue punches. Beaver TrephineBlades are handcrafted with a sharp, uniform cutting edge, designed for a precise, symmetrical cut of the corneal layers. 8.0 mm diameter
Catalog No. NC1312079 Supplier Beaver Visitec Intl Ltd Supplier No. 0009713
Beaver-Visitec o ers a comprehensive selection of corneal trephine blades, handles and tissue punches. Beaver TrephineBlades are handcrafted with a sharp, uniform cutting edge, designed for a precise, symmetrical cut of the corneal layers. 8.0 mm diameter