Learn More
Chemglass Life Sciences Valve, Chem-Vac, Glass Portion Only, 180°, 0-8mm Bore, 19mm OD Barrel, 12.7mm X 1.6W Sidearm
SDP
Catalog No. CG90402
Encompass_Preferred
Valve, Chem-Vac, Glass Portion Only, 180¿, 0-8mm Bore, 19mm OD Barrel, 12.7mm X 1.6W Sidearm. CHEM-VAC valve glass portion only. Valve has two sidearms sealed to the barrel at a 180¿ angle to one another and on the same plane. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed.
Catalog No. CG90402 Supplier Chemglass Life Sciences Supplier No. CG90402
Valve, Chem-Vac, Glass Portion Only, 180¿, 0-8mm Bore, 19mm OD Barrel, 12.7mm X 1.6W Sidearm. CHEM-VAC valve glass portion only. Valve has two sidearms sealed to the barrel at a 180¿ angle to one another and on the same plane. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed.