Learn More
C&G Containers 8OZ AMB BOST RND BTL LV3 12/CS
SDP
Catalog No. NC9462848
Encompass_Preferred
8oz Amber glass Boston Round 24-400 Black Phenolic Polycone (assembled only, bottle with cap attached) L3 12/cs
Catalog No. NC9462848 Supplier C&G Containers Supplier No. LAB0089011
8oz Amber glass Boston Round 24-400 Black Phenolic Polycone (assembled only, bottle with cap attached) L3 12/cs