Learn More
Alkali Scientific 8oz. Drosophila Bottles Round
SDP
Catalog No. 503026559
Encompass_Preferred
These 8oz Drosophila Square Bottom Bottles are specifically designed for fruit fly (Drosophila) culture applications in research laboratory and educational environments Constructed from high-quality polypropylene these bottles offer superior durability making them resistant to cracking and warping while remaining lightweight for easy handling The autoclavable design allows for high-temperature sterilization ensuring a contamination-free environment for reliable and reproducible experiments The square-bottom design provides enhanced stability reducing the risk of tipping and making them easy to stack and store A bottom indent is incorporated to help secure food in place ensuring a controlled feeding environment that supports optimal culture development The bottles measure 60x130mm providing ample space for fruit fly colonies while allowing for proper airflow The bottle openings are designed to accept standard-size paper lids foam plugs or cotton balls offering flexibility in seal
Catalog No. 50-302-6559 Supplier Alkali Scientific Supplier No. DB306
These 8oz Drosophila Square Bottom Bottles are specifically designed for fruit fly (Drosophila) culture applications in research laboratory and educational environments Constructed from high-quality polypropylene these bottles offer superior durability making them resistant to cracking and warping while remaining lightweight for easy handling The autoclavable design allows for high-temperature sterilization ensuring a contamination-free environment for reliable and reproducible experiments The square-bottom design provides enhanced stability reducing the risk of tipping and making them easy to stack and store A bottom indent is incorporated to help secure food in place ensuring a controlled feeding environment that supports optimal culture development The bottles measure 60x130mm providing ample space for fruit fly colonies while allowing for proper airflow The bottle openings are designed to accept standard-size paper lids foam plugs or cotton balls offering flexibility in seal