Learn More
C&G Containers 8 Oz Glass Jar, PTFE Lined Cap, Wide Mouth Design, 8 oz
SDP
Catalog No. NC1205352
Encompass_Preferred
The 8 oz lab-grade clear glass jar with a 58-400 continuous thread closure is ideal for secure containment of laboratory samples and chemicals. Its PTFE-lined closure offers superior chemical resistance and a low moisture transmission rate, ensuring the integrity of contents. The wide mouth design facilitates easy filling and dispensing, making it perfect for high purity chemicals, solvents, and diagnostics. Manufactured in the USA, it combines practicality with enhanced chemical safeguarding, setting it apart from other containers.
Catalog No. NC1205352 Supplier C&G Containers Supplier No. LFM008308200
The 8 oz lab-grade clear glass jar with a 58-400 continuous thread closure is ideal for secure containment of laboratory samples and chemicals. Its PTFE-lined closure offers superior chemical resistance and a low moisture transmission rate, ensuring the integrity of contents. The wide mouth design facilitates easy filling and dispensing, making it perfect for high purity chemicals, solvents, and diagnostics. Manufactured in the USA, it combines practicality with enhanced chemical safeguarding, setting it apart from other containers.