Learn More
Grainger Freeze Pack 4 in L 6-1/2 in W PK18
Catalog No. NC1413638
Encompass_Preferred
Reusable, Colder than Ice, Safe Non-Toxic. Featuring nontoxic refrigerants, these hermetically sealed, leakproof freeze packs can stay cold for days. They keep perishables and pharmaceuticals refrigerated during shipping.
Catalog No. NC1413638 Supplier Grainger Supplier No. 4YRJ3
Reusable, Colder than Ice, Safe Non-Toxic. Featuring nontoxic refrigerants, these hermetically sealed, leakproof freeze packs can stay cold for days. They keep perishables and pharmaceuticals refrigerated during shipping.