Learn More
Perkin Elmer US LLC Snoop Liquid Leak Detector, 8 oz.
Catalog No. 509045815
Encompass_Preferred
The Snoop solution is composed of deionized water and a surfactant that produces sustained bubbling when it comes into contact with gas leaks. The solution is non-toxic, non-corrosive, non-flammable and does not contain chlorine. Other Features: Flexible snooper tube permits access to hard-to-reach areas Formula contains a corrosion inhibitor for added protection Solution dries clean, without stainingNote that this equivalent to PerkinElmer part number 1696000.
Catalog No. 50-904-5815 Supplier Perkin Elmer US LLC Supplier No. N9301329
The Snoop solution is composed of deionized water and a surfactant that produces sustained bubbling when it comes into contact with gas leaks. The solution is non-toxic, non-corrosive, non-flammable and does not contain chlorine. Other Features: Flexible snooper tube permits access to hard-to-reach areas Formula contains a corrosion inhibitor for added protection Solution dries clean, without stainingNote that this equivalent to PerkinElmer part number 1696000.