Learn More
C&G Containers C&G Containers 8 Oz Glass Jar, PTFE Lined Screw Cap, Airtight Seal, 96/Case
SDP
Catalog No. NC1997392
Encompass_Preferred
The C & G Containers 8 oz Straight-Sided Clear Glass Jar is designed for optimal laboratory and chemical storage. Featuring a PTFE-lined cap for exceptional chemical resistance, this jar ensures secure containment for sensitive materials. Its straight-sided design allows for easy filling, access, and labeling, making it an ideal choice for various scientific applications. With a high capacity of 240 mL and airtight seal, it stands out in the market for reliable, clean storage solutions.
Catalog No. NC1997392 Supplier C&G Containers Supplier No. LFA008900000
The C & G Containers 8 oz Straight-Sided Clear Glass Jar is designed for optimal laboratory and chemical storage. Featuring a PTFE-lined cap for exceptional chemical resistance, this jar ensures secure containment for sensitive materials. Its straight-sided design allows for easy filling, access, and labeling, making it an ideal choice for various scientific applications. With a high capacity of 240 mL and airtight seal, it stands out in the market for reliable, clean storage solutions.