Learn More
MSC Guardian Equipment Aquasep Eyewash Preservative, Bacteriostatic, 8 oz, 4 Bottles
Catalog No. NC1545633
Encompass
The Guardian G1540BA-R Aquasep is a bacteriostatic water preservative additive for portable eyewash stations. It is designed to protect against the growth of bacteria and other impurities in portable units, where water may sit in the tank between scheduled refills and maintenance checks.
  • Protects against the growth of bacteria, fungus, algae, and acanthamoeba
  • Each case includes four 8 oz (236 mL) bottles
  • One bottle treats up to 20 gal. of clean potable water
  • Helps preserve flushing fluid for up to 6 months
  • Designed for self-contained eyewash units
  • Concentrate that must be diluted with potable water
This item is not returnable. View return policy
Catalog No. NC1545633 Supplier MSC Supplier No. 86205325
The Guardian G1540BA-R Aquasep is a bacteriostatic water preservative additive for portable eyewash stations. It is designed to protect against the growth of bacteria and other impurities in portable units, where water may sit in the tank between scheduled refills and maintenance checks.
  • Protects against the growth of bacteria, fungus, algae, and acanthamoeba
  • Each case includes four 8 oz (236 mL) bottles
  • One bottle treats up to 20 gal. of clean potable water
  • Helps preserve flushing fluid for up to 6 months
  • Designed for self-contained eyewash units
  • Concentrate that must be diluted with potable water