Learn More
US Plastic Corporation 8.3 oz. Wide Mouth Round Natural HDPE Jar with 53/400 White Ribbed Cap with F217 Liner
SDP
Catalog No. NC3455949
Encompass
These wide mouth round jars are great for samples and can be used for liquids, pastes, and powders.HDPE jarPolypropylene cap with F217 linerDimensions: 2.64" Dia. x 4" Hgt.On thread: 2.05" Dia.Corrosion resistantLighter than glassMeets FDA standards
Catalog No. NC3455949 Supplier US Plastic Corporation Supplier No. 66415
These wide mouth round jars are great for samples and can be used for liquids, pastes, and powders.HDPE jarPolypropylene cap with F217 linerDimensions: 2.64" Dia. x 4" Hgt.On thread: 2.05" Dia.Corrosion resistantLighter than glassMeets FDA standards