Learn More
MSC Dynalon Labware Polyethylene Jar, Wide Mouth, 8 oz, Autoclavable, 6/Pack
Catalog No. 502241188
Encompass
This polyethylene jar is engineered for laboratory applications, providing an 8 oz capacity and a wide mouth for convenient filling and dispensing. Its autoclavable design ensures sterility, making it suitable for various scientific uses.
  • Autoclavable for sterilization
  • Includes closure
  • Round shape with a wide mouth
  • Constructed from polyethylene
  • White body color with translucent appearance
  • Diameter of 3.5 in
  • Height of 66 mm
This item is not returnable. View return policy
Catalog No. 50-224-1188 Supplier MSC Supplier No. 65361172
This polyethylene jar is engineered for laboratory applications, providing an 8 oz capacity and a wide mouth for convenient filling and dispensing. Its autoclavable design ensures sterility, making it suitable for various scientific uses.
  • Autoclavable for sterilization
  • Includes closure
  • Round shape with a wide mouth
  • Constructed from polyethylene
  • White body color with translucent appearance
  • Diameter of 3.5 in
  • Height of 66 mm