Learn More
Cayman Chemical 8pIpRIdIno NAD sodIM sal 700ug
SDP
Catalog No. 502917243
Encompass_Preferred
A derivative of NAD+; has been used in the determination of PARP target proteins; a synthetic intermediate in the synthesis of cADPR derivatives
Catalog No. 50-291-7243 Supplier Cayman Chemical Supplier No. 38487700 UG
A derivative of NAD+; has been used in the determination of PARP target proteins; a synthetic intermediate in the synthesis of cADPR derivatives