Learn More
USA Scientific Inc 8-pl upright freezer rk
Catalog No. 502745099
Encompass
8-place (4 long x 2 high) upright freezer rack for 4 3/4 in H boxes, 24 in L x 5 7/8 in W x 9 7/8 in H, stainless steel
Catalog No. 50-274-5099 Supplier USA Scientific Inc Supplier No. 26239008
8-place (4 long x 2 high) upright freezer rack for 4 3/4 in H boxes, 24 in L x 5 7/8 in W x 9 7/8 in H, stainless steel