Learn More
USA Scientific Inc 8-place chest freezer rk
Catalog No. 502744886
Encompass
8-place (1 long x 8 high) chest freezer rack for 3 in H boxes, 5 1/2 L x 5 5/8 W x 25 7/8 H, stainless steel
Catalog No. 50-274-4886 Supplier USA Scientific Inc Supplier No. 26038000
8-place (1 long x 8 high) chest freezer rack for 3 in H boxes, 5 1/2 L x 5 5/8 W x 25 7/8 H, stainless steel