Learn More
Perkin Elmer US LLC 8-Position Turntable with High Pressure 100 mL, 100 Bar Digestion Vessels, with DPC, Complete Assembly
Catalog No. 502111670
Encompass_Preferred
This assembly includes: 8 High Pressure 100 mL (100 Bar) Digestion Vessels made of TFM with TFM Sample Vessel Insert Ceramic Pressure Jacket Rupture Disc Vessel cap (with DPC) 8-Position Turntable Gas Containment Manifold te:
Catalog No. 50-211-1670 Supplier Perkin Elmer US LLC Supplier No. N3131011
Add to Cart
Add to Cart
Request a Quote
This assembly includes: 8 High Pressure 100 mL (100 Bar) Digestion Vessels made of TFM with TFM Sample Vessel Insert Ceramic Pressure Jacket Rupture Disc Vessel cap (with DPC) 8-Position Turntable Gas Containment Manifold te: