Learn More
Medchemexpress LLC 8PPD-Q | 99.9% | 326.43 | 25 MG
SDP
Catalog No. 5000384204
Encompass_Preferred
8PPD-Q, a para-phenylenediamine quinone, is a rubber antioxidant. It exhibits superior antioxidant and anti-ozone activities but is toxic to aquatic species, particularly salmonids. 8PPD-Q can be utilized for environmental pollution research.
  • Rubber antioxidant
  • Superior antioxidant activity
  • Anti-ozone activity
  • Can be used for environmental pollution research
Catalog No. 50-003-84204 Supplier Medchemexpress LLC Supplier No. 25MGHY171702
8PPD-Q, a para-phenylenediamine quinone, is a rubber antioxidant. It exhibits superior antioxidant and anti-ozone activities but is toxic to aquatic species, particularly salmonids. 8PPD-Q can be utilized for environmental pollution research.
  • Rubber antioxidant
  • Superior antioxidant activity
  • Anti-ozone activity
  • Can be used for environmental pollution research