Learn More
Medchemexpress LLC 8PPD-Q | 99.9% | 326.43 | 5 MG
SDP
Catalog No. 5000386511
Encompass_Preferred
8PPD-Q, a para-phenylenediamine quinone, is a rubber antioxidant. It has superior antioxidant and anti-ozone activities but is toxic to the aquatic species, especially salmonids. 8PPD-Q can be used for environmental pollution research.
  • Functions as a para-phenylenediamine quinone compound.
  • Acts as a rubber antioxidant.
  • Exhibits superior antioxidant activities.
  • Provides superior anti-ozone activities.
  • Applicable for environmental pollution research.
Catalog No. 50-003-86511 Supplier Medchemexpress LLC Supplier No. HY1717025MG
8PPD-Q, a para-phenylenediamine quinone, is a rubber antioxidant. It has superior antioxidant and anti-ozone activities but is toxic to the aquatic species, especially salmonids. 8PPD-Q can be used for environmental pollution research.
  • Functions as a para-phenylenediamine quinone compound.
  • Acts as a rubber antioxidant.
  • Exhibits superior antioxidant activities.
  • Provides superior anti-ozone activities.
  • Applicable for environmental pollution research.