Learn More
Medchemexpress LLC (8R,9S)-Talazoparib ((8R,9S)-BMN-673) | 1207456-00-5 | 98.3% | 380.35 | 100 MG
SDP
Catalog No. 5000395095
Encompass_Preferred
(8R,9S)-Talazoparib is an enantiomer of Talazoparib and acts as a PARP1 inhibitor with an IC50 of 144 nM. This product is for research use only.
  • Acts as a PARP-1 inhibitor with an IC50 of 144 nM.
  • Exhibits an EC50 of 1135 nM for cytotoxicity against BRCA2-deficient human Capan1 cells.
  • Shows an EC50 of 864 nM for inhibition of PARP in human LoVo cells.
  • Demonstrates a GI50 of 1807 nM for potentiation of temozolomide-induced cytotoxicity in human LoVo cells.
  • Displays an EC50 of 864 nM in cellular PARylation assay.
  • Shows an EC50 of 1135 nM for Capan-1 cells (BRCA2 mutant).
Catalog No. 50-003-95095 Supplier Medchemexpress LLC Supplier No. HY16106A100MG
(8R,9S)-Talazoparib is an enantiomer of Talazoparib and acts as a PARP1 inhibitor with an IC50 of 144 nM. This product is for research use only.
  • Acts as a PARP-1 inhibitor with an IC50 of 144 nM.
  • Exhibits an EC50 of 1135 nM for cytotoxicity against BRCA2-deficient human Capan1 cells.
  • Shows an EC50 of 864 nM for inhibition of PARP in human LoVo cells.
  • Demonstrates a GI50 of 1807 nM for potentiation of temozolomide-induced cytotoxicity in human LoVo cells.
  • Displays an EC50 of 864 nM in cellular PARylation assay.
  • Shows an EC50 of 1135 nM for Capan-1 cells (BRCA2 mutant).