Learn More
Chemglass Life Sciences 8-Strip PCR Caps, Domed, Non-Sterile, 125/Pk
SDP
Catalog No. 501897741
Encompass_Preferred
Compatible with both NEST PCR 8-strip tube and PCR plate.USP grade VI PolypropyleneTight sealing, easy to openDomed cap helps prevent liquid evaporationAutoclavable
Catalog No. 50-189-7741 Supplier Chemglass Life Sciences Supplier No. CGN3610D01
Compatible with both NEST PCR 8-strip tube and PCR plate.USP grade VI PolypropyleneTight sealing, easy to openDomed cap helps prevent liquid evaporationAutoclavable