Learn More
Chemglass Life Sciences 8-Strip PCR Caps, Flat, Non-Sterile, 125/Pk
SDP
Catalog No. 501897740
Encompass_Preferred
Compatible with both NEST PCR 8-strip tube and PCR plate.USP grade VI PolypropyleneTight sealing, easy to openFlat cap is ideal for qPCRAutoclavable
Catalog No. 50-189-7740 Supplier Chemglass Life Sciences Supplier No. CGN3610F01
Compatible with both NEST PCR 8-strip tube and PCR plate.USP grade VI PolypropyleneTight sealing, easy to openFlat cap is ideal for qPCRAutoclavable