Learn More
Genesee Scientific Corporation Olympus 8-Strip PCR Tubes, 0.2 mL Volume, RNase-Free, Made in USA
SDP
Catalog No. NC2293462
Encompass
The Olympus 8-Strip PCR Tubes with Caps are designed for efficient PCR applications. Made from virgin, FDA-approved polypropylene, these tubes ensure optimal heat transfer with ultra-thin walls. Each strip features individually attached flat caps with a 45º hinge for easy access. Certified free of contaminants, they enhance handling and maintain integrity during sensitive assays. The 120-pack offers convenience for laboratory use, making them ideal for consistent and reliable results in molecular biology.
This item is not returnable. View return policy
Catalog No. NC2293462 Supplier Genesee Scientific Corporation Supplier No. 27125UA
The Olympus 8-Strip PCR Tubes with Caps are designed for efficient PCR applications. Made from virgin, FDA-approved polypropylene, these tubes ensure optimal heat transfer with ultra-thin walls. Each strip features individually attached flat caps with a 45º hinge for easy access. Certified free of contaminants, they enhance handling and maintain integrity during sensitive assays. The 120-pack offers convenience for laboratory use, making them ideal for consistent and reliable results in molecular biology.