Learn More
Edge Biosystems Inc Optima DTR 8-Well Strip Kit
SDP
Supplier:  Edge Biosystems Inc 8WELL1 OPTIMA DTR 8 WELL
Encompass
Optima DTR employs a new, more cost-effective resin than Performa DTR, provides the same high-quality results at a lower price, and is available in 8-well, 96-well, and 384-well formats.
Catalog No. NC1269358