Learn More
Cellvis 8WELL CHAMBERED COVER GLASS #1
Catalog No. NC1862490
Encompass
8 Well Chambered Cover Glass with #1 cover glass (0.13-0.16mm). Individually packed, with lid, Sterilized. Designed for high resolution imaging such as confocal microscopy.•Suitable for long term tissue culture•Manufactured in a class 100,000 clean room•Frame made from virgin polystyrene.•German high quality cover glass of superior optical quality, glass thickness is 0.170±0.005mm •A USP class VI adhesive is used to assemble the cover glass and the plate.•Sterilized by Gamma radiation.
Catalog No. NC1862490 Supplier Cellvis Supplier No. C81N
8 Well Chambered Cover Glass with #1 cover glass (0.13-0.16mm). Individually packed, with lid, Sterilized. Designed for high resolution imaging such as confocal microscopy.•Suitable for long term tissue culture•Manufactured in a class 100,000 clean room•Frame made from virgin polystyrene.•German high quality cover glass of superior optical quality, glass thickness is 0.170±0.005mm •A USP class VI adhesive is used to assemble the cover glass and the plate.•Sterilized by Gamma radiation.