Learn More
Ibidi USA 8 Well Chamber, removable
SDP
Catalog No. NC1535706
Encompass
The 8 Well Chamber, removable is a removable silicone chamber, mounted on a glass slide, for cell culture and immunofluorescence staining. It allows the use of standard cultivation, staining and mounting techniques with coverslip sealing. After mounting the glass slide with a coverslip, it is ready for the use with upright and inverted microscopes, as well as for long term storage. Suitable 24 mm×60 mm coverslips are provided by ibidi (Cat.# 10811). Glass slide, silicone chamber, sterile, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC1535706 Supplier Ibidi USA Supplier No. 80841
The 8 Well Chamber, removable is a removable silicone chamber, mounted on a glass slide, for cell culture and immunofluorescence staining. It allows the use of standard cultivation, staining and mounting techniques with coverslip sealing. After mounting the glass slide with a coverslip, it is ready for the use with upright and inverted microscopes, as well as for long term storage. Suitable 24 mm×60 mm coverslips are provided by ibidi (Cat.# 10811). Glass slide, silicone chamber, sterile, 15 pcs./box