Learn More
Fuji Medical Systems Green Fuji Super HR-U X-Ray Film 8 x 10 in
Catalog No. NC1110786
Encompass
Super HR-U is a high contrast green sensitive x-ray film for use with green-emitting rare earth screens. The New all-around x-ray film for general applications.
Catalog No. NC1110786 Supplier Fuji Medical Systems Supplier No. 16082189
Super HR-U is a high contrast green sensitive x-ray film for use with green-emitting rare earth screens. The New all-around x-ray film for general applications.