Learn More
Cole-Parmer Bags Ldpe 8x10 500/pk, Clear - Polyethylene
Supplier:  Cole-Parmer 0650514
Encompass_Preferred
Bags Ldpe 8x10 500/pk, Clear - Polyethylene. Specifications: Product Type Low-Density Bags, Material Polyethylene, Size 8" W x 10" L, Thickness (mil) 3, Color Clear.
Catalog No. NC9637198