Learn More
Analytical Sales & Services 8x3mm PTFE, Magnetic Stir Bar with Pivot Ring
Catalog No. 5000807250
Encompass
8x3mm PTFE, Magnetic Stir Bar with Pivot Ring\n\n• Diameter: 3mm\n• Length: 8mm\n\nQty/Pkg: 10\n
Catalog No. 50-008-07250 Supplier Analytical Sales & Services Supplier No. 80310
8x3mm PTFE, Magnetic Stir Bar with Pivot Ring\n\n• Diameter: 3mm\n• Length: 8mm\n\nQty/Pkg: 10\n