Learn More
Thomas Scientific Glass Fiber Pad, 9 cm,
SDP
Catalog No. NC0929079
Encompass
Pads are compatible with CEM, other microwave units and infrared analysis such as Arizona Instr., Sartorius, Ohaus and Mettler. 9-cm diameter in diameter and meet AOAC criteria for conducting analysis. Packaged in packs of 200.
Catalog No. NC0929079 Supplier Thomas Scientific Supplier No. 9885E25
Pads are compatible with CEM, other microwave units and infrared analysis such as Arizona Instr., Sartorius, Ohaus and Mettler. 9-cm diameter in diameter and meet AOAC criteria for conducting analysis. Packaged in packs of 200.