Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. B5054200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5054200MG 200 mg
B50541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5054200MG Supplier TCI America Supplier No. B5054200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1533406-38-0
Molecular Formula C30H28BNO2
Molecular Weight (g/mol) 445.369
MDL Number MFCD28130384
InChI Key VQLXKJLKLNGTQF-UHFFFAOYSA-N
Synonym 9-(3-Biphenylyl)carbazole-3-boronic Acid Pinacol Ester, 2-[9-([1,1′C-Biphenyl]-3-yl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 90336757
IUPAC Name 9-(3-phenylphenyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=CC=CC=C43)C5=CC=CC(=C5)C6=CC=CC=C6

Specifications

Melting Point 238°C
Color White
Quantity 200 mg
Formula Weight 445.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-([1,1′-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.