Learn More
MOLDPRO INC 1.2 mL AA Sample Cup, 9 X 25 mm, Clear, 2000/pkg, rounded skirted bottom, PP Copolymer
Catalog No. NC1696307
Encompass_Preferred
MoldPro AA Sample Cups are standard size and fit systems by Cetac, PerkinElmer, Agilent/Varian, Shimadzu and SEAL • Made from virgin polypropylene • Excellent visibility • Good resistance to acids • MP-106 1000 tubes/pack, MP-107 2000/pack
Catalog No. NC1696307 Supplier MOLDPRO INC Supplier No. MP107
MoldPro AA Sample Cups are standard size and fit systems by Cetac, PerkinElmer, Agilent/Varian, Shimadzu and SEAL • Made from virgin polypropylene • Excellent visibility • Good resistance to acids • MP-106 1000 tubes/pack, MP-107 2000/pack