Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10,12,13-Tetrabromostearic Acid 97.0+%, TCI America™
SDP

Catalog No. T004710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T004710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T004710G Supplier TCI America Supplier No. T004710G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1794-89-4
Molecular Formula C18H32Br4O2
Molecular Weight (g/mol) 600.068
MDL Number MFCD00021796
InChI Key HTORNZPNCYXGMR-UHFFFAOYSA-N
Synonym 9,10,12,13-Tetrabromooctadecanoic Acid
PubChem CID 98942
IUPAC Name 9,10,12,13-tetrabromooctadecanoic acid
SMILES CCCCCC(C(CC(C(CCCCCCCC(=O)O)Br)Br)Br)Br

Specifications

Melting Point 115°C
Color White
Quantity 10 g
Formula Weight 600.07
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9,10,12,13-Tetrabromostearic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.