Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Anthracenedicarboxaldehyde 80%, Thermo Scientific™

Catalog No. AC346790050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC346790050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC346790050 Supplier Thermo Scientific Chemicals Supplier No. 346790050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7044-91-9
Molecular Formula C16H10O2
Molecular Weight (g/mol) 234.25
MDL Number MFCD00045386
InChI Key SBRUFOSORMQHES-UHFFFAOYSA-N
Synonym 9,10-anthracenedicarboxaldehyde, anthracene-9,10-dicarboxaldehyde, anthracene-9,10-dialdehyde, 9,10-diformylanthracene, pubchem15118, 9,10-anthracenedicarbaldehyde, 9,10-anthracenedicarbaldehyde #, 9,10-anthracene dicarboxyaldehyde
PubChem CID 81500
ChEBI CHEBI:51295
IUPAC Name anthracene-9,10-dicarbaldehyde
SMILES O=CC1=C2C=CC=CC2=C(C=O)C2=CC=CC=C12

Specifications

CAS Min % 75.0
CAS Max % 100.0
Color Orange to Brown
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 234.25
Percent Purity ≥75%
Physical Form Crystalline Powder
Chemical Name or Material 9, 10-Anthracenedicarboxaldehyde, 80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.