Learn More
Medchemexpress LLC 9,10-Anthracenedione, 1,4-dihydroxy- | 81-64-1 | 98.0% | 2.4 MG
SDP
Catalog No. 5000410285
Encompass_Preferred
Quinizarin (1,4-Dihydroxyanthraquinone) is a chemical compound that interacts with DNA through an intercalating mode, with a Kd of 86.1 μM. It is considered part of anticancer agents like Doxorubicin, Daunorubicin, and Adriamycin. Quinizarin is also utilized as a fungicide and pesticide chemical and has demonstrated the capacity to inhibit tumor cell growth.
  • Interacts with DNA by intercalating mode (Kd=86.1 μM).
  • Used as a fungicide and pesticide chemical.
  • Shown to inhibit tumor cell growth.
  • Categorized as a fluorescent dye.
Catalog No. 50-004-10285 Supplier Medchemexpress LLC Supplier No. HYD022610MM/1ML
Quinizarin (1,4-Dihydroxyanthraquinone) is a chemical compound that interacts with DNA through an intercalating mode, with a Kd of 86.1 μM. It is considered part of anticancer agents like Doxorubicin, Daunorubicin, and Adriamycin. Quinizarin is also utilized as a fungicide and pesticide chemical and has demonstrated the capacity to inhibit tumor cell growth.
  • Interacts with DNA by intercalating mode (Kd=86.1 μM).
  • Used as a fungicide and pesticide chemical.
  • Shown to inhibit tumor cell growth.
  • Categorized as a fluorescent dye.